Human Recombinant FGF-18 (from E. coli)
Supplier: Peprotech
AF-100-28-5UG AF-100-28-25UG AF-100-28-100UG AF-100-28-250UG AF-100-28-500UG AF-100-28-1MG
10778-082EA 289.23 CAD
10778-082 10778-084 10778-086 10778-088 10778-090 10778-092
Human Recombinant FGF-18 (from E. coli)
Proteins and Peptides
FGF-18 is a heparin-binding growth factor that belongs to the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-18 is an essential regulator of long bone and calvarial development. FGF-18 signals through FGFR 1c, 2c, 3c, and 4. Recombinant Human FGF-18 derived from
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.