Human Recombinant FGF-10 (from E. coli)
Supplier: Peprotech
AF-100-26-5UG AF-100-26-25UG AF-100-26-100UG AF-100-26-250UG AF-100-26-500UG AF-100-26-1MG
10778-070EA 300.99 CAD
10778-070 10778-072 10778-074 10778-076 10778-078 10778-080
Human Recombinant FGF-10 (from E. coli)
Proteins and Peptides
FGF-10 is a heparin-binding growth factor that belongs to the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-10 is most related to KGF/FGF-7, and is expressed during the development and, preferentially, in adult lungs. It signals through the FGFR 2b. Recombinant Human FGF-10 is a 19.3 kDa protein consisting of 170 amino acid residues.
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.