Human Recombinant KGF (from E. coli)
Supplier: Peprotech
AF-100-19-2UG AF-100-19-10UG AF-100-19-50UG AF-100-19-100UG AF-100-19-250UG AF-100-19-500UG AF-100-19-1MG
10777-994EA 288.29 CAD
10777-994 10777-996 10777-998 10778-000 10778-002 10778-004 10778-006
Human Recombinant KGF (from E. coli)
Proteins and Peptides
KGF (FGF-7) is one of 23 known members of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth, and the regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. KGF (FGF-7) is a mitogen factor specific for epithelial cells and keratinocytes. KGF (FGF-7) signals through FGFR 2b. KGF (FGF-7) plays a role in kidney and lung development, as well as in angiogenesis and wound healing. Recombinant Human KGF (FGF-7) is an 18.9 kDa protein consisting of 163 amino acid residues.
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.