Human Recombinant FGF-basic (from E. coli)
Supplier: Peprotech
AF-100-18C-100UG AF-100-18C-250UG AF-100-18C-500UG AF-100-18C-1MG
10777-986EA 780.44 CAD
10777-986 10777-988 10777-990 10777-992
Human Recombinant FGF-basic (from E. coli)
Proteins and Peptides
FGF-basic is one of 23 known members of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-basic is a non-glycosylated, heparin-binding growth factor that is expressed in the brain, pituitary, kidney, retina, bone, testis, adrenal gland, liver, monocytes, epithelial cells and endothelial cells. FGF-basic signals through FGFR 1b, 1c, 2c, 3c and 4. Recombinant Human FGF-basic is a 16.4 kDa protein consisting of 146 amino acid residues.
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.