Human Recombinant PLGF-I (from E. coli)
Supplier: Peprotech
AF-100-06-5UG AF-100-06-25UG AF-100-06-100UG AF-100-06-250UG AF-100-06-500UG AF-100-06-1MG
10777-958EA 304.45 CAD
10777-958 10777-960 10777-962 10777-964 10777-966 10777-968
Human Recombinant PLGF-I (from E. coli)
Proteins and Peptides
PlGF-1 is an angiogenic factor that belongs to the cysteine-knot superfamily of growth factors. PlGF-1 is expressed in placental tissues, the colon and mammary carcinomas. It signals through the VEGFR-1/FLT1 receptor, and stimulates endothelial cell proliferation and migration. Recombinant Human PlGF-1 is a 29.7 kDa disulfide-linked homodimeric protein of two 132 amino acid polypeptide chains.Manufactured using all Animal-Free reagents.
  • Carrier free
  • Animal-free
MLPAVPPQQW ALSAGNGSSE VEVVPFQEVW GRSYCRALER LVDVVSEYPS EVEHMFSPSC VSLLRCTGCC GDENLHCVPV ETANVTMQLL KIRSGDRPSY VELTFSQHVR CECRPLREKM KPERCGDAVP RR
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.