Mouse Recombinant gAcrp30 (from E. coli)
Supplier: Peprotech
450-27-5UG 450-27-25UG 450-27-50UG 450-27-100UG 450-27-250UG 450-27-500UG 450-27-1MG
10774-712EA 209.84 CAD
10774-712 10774-714 10774-716 10774-718 10774-720 10774-722 10774-724
Mouse Recombinant gAcrp30 (from E. coli)
Proteins and Peptides
gAcrp30 is a naturally occurring globular protein obtained by proteolytic processing of adiponectin. Adiponectin is produced and secreted exclusively by adipocytes, and is a relatively abundant plasma protein, accounting for up to 0.05% of total serum protein. Like Adiponectin, gAcrp30 is capable of decreasing hyperglycemia and reversing insulin resistance. Additionally, gAcrp30 has been shown to be an important factor in promoting fat loss by signaling muscle to absorb and burn Free-Fatty Acids (FFAs). The signaling receptors for adiponectin and gAcrp30 have recently been identified and named AdipoR1 and AdipoR2. AdipoR2 is predominantly expressed in the liver. Recombinant Murine gAcrp30 is a 16.6 kDa protein consisting of 145 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.