Human Recombinant RELMbeta (from E. coli)
Supplier: Peprotech
450-22-5UG 450-22-25UG 450-22-100UG 450-22-250UG 450-22-500UG 450-22-1MG
10774-652EA 209.84 CAD
10774-652 10774-654 10774-656 10774-658 10774-660 10774-662
Human Recombinant RELMbeta (from E. coli)
Proteins and Peptides
RELMβ (Resistin-like molecule β/FIZZ2) is a disulfide-linked, homodimeric protein expressed in the epithelium of the colon and small bowel. The biological functions of RELMβ, and its molecular targets, are not fully known, but it has been suggested that it plays a regulatory role during inflammation, and may also act to establish links among adipose tissue, the intestine and the liver. Interestingly, the molecular structure of RELMβ is highly homologous to that of the adipose-derived cytokines, resistin and RELMα. These proteins share a highly conserved C-terminal domain, characterized by 10 cysteine residues with a unique spacing motif of C-X11-C-X8-C-X-C-X3-C-X10-C-X-C-X-C-X9-C-C. Recombinant Human RELMβ is a 19.0 kDa protein, consisting of two identical 89 amino acid polypeptide chains linked by a single disulfide bond.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.