Rat Recombinant EGF (from E. coli)
Supplier: Peprotech
400-25-20UG 400-25-100UG 400-25-250UG 400-25-500UG 400-25-1MG
10774-602EA 221.03 CAD
10774-602 10774-604 10774-606 10774-608 10774-610
Rat Recombinant EGF (from E. coli)
Proteins and Peptides
EGF is a potent growth factor that stimulates the proliferation of various epidermal and epithelial cells. Additionally, EGF has been shown to inhibit gastric secretion, and to be involved in wound healing. EGF signals through a receptor known as c-erbB, which is a class I tyrosine kinase receptor. This receptor also binds with TGF-α and VGF (vaccinia virus growth factor). Recombinant Rat EGF is a 6.2 kDa globular protein containing 54 amino acid residues, including 3 intramolecular disulfide bonds.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.