Rat Recombinant GM-CSF (from E. coli)
Supplier: Peprotech
400-23-5UG 400-23-20UG 400-23-50UG 400-23-100UG 400-23-250UG 400-23-500UG 400-23-1MG
10774-576EA 221.44 CAD
10774-576 10774-578 10774-580 10774-582 10774-584 10774-586 10774-588
Rat Recombinant GM-CSF (from E. coli)
Proteins and Peptides
GM-CSF is a hematopoietic growth factor that stimulates the development of neutrophils and macrophages, and promotes the proliferation and development of early erythroid megakaryocytic and eosinophilic progenitor cells. It is produced in by endothelial cells, monocytes, fibroblasts and T lymphocytes. GM-CSF inhibits neutrophil migration and enhances the functional activity of the mature end-cells. The human and murine molecules are species-specific and exhibit no cross-species reactivity. Recombinant Rat GM-CSF is a 14.5 kDa globular protein consisting of 128 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.