Rat Recombinant IFN-Gamma (from E. coli)
Supplier: Peprotech
400-20-20UG 400-20-100UG 400-20-250UG 400-20-500UG 400-20-1MG
10774-548EA 226.59 CAD
10774-548 10774-550 10774-552 10774-554 10774-556
Rat Recombinant IFN-Gamma (from E. coli)
Proteins and Peptides
IFN-γ is an acid-labile interferon produced by CD4 and CD8 T lymphocytes, as well as activated NK cells. IFN-γ receptors are present in most immune cells, which respond to IFN-γ signaling by increasing the surface expression of class I MHC proteins. This promotes the presentation of antigen to T-helper (CD4+) cells. IFN-γ signaling in antigen-presenting cells, and antigen-recognizing B and T lymphocytes, regulates the antigen-specific phases of the immune response. Additionally, IFN-γ stimulates a number of lymphoid cell functions, including the anti-microbial and anti-tumor responses of macrophages, NK cells, and neutrophils. Human IFN-γ is species-specific and is biologically active only in human and primate cells. Recombinant Rat IFN-γ is a 15.6 kDa protein containing 135 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.