Human Recombinant ApoE3 (from E. coli)
Supplier: Peprotech
350-02-100UG 350-02-500UG
10774-404EA 225.8 CAD
10774-404 10774-406 10774-408
Human Recombinant ApoE3 (from E. coli)
Proteins and Peptides
ApoE belongs to a group of proteins that bind reversibly with lipoprotein and play an important role in lipid metabolism. In addition to facilitating solubilization of lipids, these proteins help to maintain the structural integrity of lipoproteins, serve as ligands for lipoprotein receptors, and regulate the activity of enzymes involved in lipid metabolism. Significant quantities of ApoE are produced in the liver and brain, and to some extent in almost every organ. ApoE is an important constituent of all plasma lipoproteins. Its interaction with specific ApoE receptor enables uptake of chylomicron remnants by liver cells, which is an essential step during normal lipid metabolism. It also binds with the LDL receptor (apo B/E). Defects in ApoE are a cause of hyperlipoproteinemia type III. ApoE exists in three major isoforms; E2, E3, and E4, which differ from one another by a single amino-acid substitution. E3 is the most common isoform and is present in 40-90% of the population. Recombinant Human ApoE3 is a 34.4 kDa protein containing 300 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.