Mouse Recombinant EG-VEGF (from E. coli)

Supplier: Peprotech

315-29-5UG 315-29-20UG 315-29-100UG 315-29-250UG 315-29-500UG 315-29-1MG
10774-392EA 239.65 CAD
10774-392 10774-394 10774-396 10774-398 10774-400 10774-402
Mouse Recombinant EG-VEGF (from E. coli)
Proteins and Peptides

EG-VEGF is a secreted angiogenetic mitogen growth factor expressed in the steroidogenic glands, ovary, testis, adrenal gland, and placenta. EG-VEGF induces proliferation, migration, and fenestration (formation of membrane discontinuities) in capillary endothelial cells derived from endocrine glands. The murine EG-VEGF gene codes for a 105 amino acid polypeptide containing an N-terminal signal sequence of 19 amino acids. Recombinant Murine EG-VEGF is a 9.6 kDa protein consisting of 86 amino acid residues, including ten cysteine residues that potentially form five pairs of intra-molecular disulfide bonds.


Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.

Order Now

SPECIFICATIONS

Protein/peptide name EG-VEGF
Environmentally Preferable
Protein synonyms Prokineticin 1|PROK1|Endocrine Gland-derived Vascular Endothelial Growth Factor
Protein/peptide type Recombinant
Protein Function Cytokines and Growth Factors
Species Mouse
Source E. coli
Conjugation Unconjugated
Gene ID 246691 Q14A28
Sequence AVITGACERD IQCGAGTCCA ISLWLRGLRL CTPLGREGEE CHPGSHKIPF LRKRQHHTCP CSPSLLCSRF PDGRYRCFRD LKNANF
Protease-free Y
Endotoxin level Low
Endotoxin content >0.1 ng/μg
Biological activity Biological Activity tests can vary. Please refer to the Product CoA for information.
Purity 0.98
Form Lyophilized
Formulation Formulation can vary. Please refer to product CoA for information.
Molecular weight 9.6 kDa
Reconstitution instructions Reconstitution instructions can vary. Please refer to the product CoA for info.
Storage conditions Store at –20 °C prior to reconstitution.
Shipping temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR