Mouse Recombinant BMP-4 (from E. coli)
Supplier: Peprotech
315-27-2UG 315-27-10UG 315-27-50UG 315-27-100UG 315-27-250UG 315-27-500UG 315-27-1MG
10774-378EA 209.84 CAD
10774-378 10774-380 10774-382 10774-384 10774-386 10774-388 10774-390
Mouse Recombinant BMP-4 (from E. coli)
Proteins and Peptides
Bone morphogenetic proteins (BMPs) constitute a subfamily within the TGF-β superfamily of structurally related signaling proteins. Members of this superfamily are widely distributed throughout the body, and are involved in diverse physiological processes during both pre- and postnatal life. Like BMP-7, BMP-4 is involved in the development and maintenance of bone and cartilage. Reduced expression of BMP-4 is associated with a number of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. PeproTech’s
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.