Mouse Recombinant Midkine (from E. coli)
Supplier: Peprotech
315-25-5UG 315-25-20UG 315-25-100UG 315-25-250UG 315-25-500UG 315-25-1MG
10774-352EA 239.65 CAD
10774-352 10774-354 10774-356 10774-358 10774-360 10774-362
Mouse Recombinant Midkine (from E. coli)
Proteins and Peptides
Midkine (MK) and its functionally-related protein pleiotrophin are heparin-binding neurotrophic factors that signal through the same receptor, known as anaplastic lymphoma kinase (ALK). MK plays an important regulatory role in epithelial-mesenchymal interactions during fetal development and in postnatal lung development. MK chemoattracts embryonic neurons, neutrophils and macrophages, and exerts angiogenic, growth and survival activities during tumorgenesis. Recombinant Murine Midkine is a 13.3 kDa protein containing 120 amino acid residues including five intra-molecular disulfide bonds.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.