Mouse Recombinant Sonic (from E. coli)
Supplier: Peprotech
315-22-250UG 315-22-500UG 315-22-1MG
10774-322EA 2620.87 CAD
10774-322 10774-324 10774-326
Mouse Recombinant Sonic (from E. coli)
Members of the Hedgehog (Hh) family are highly conserved proteins that are widely represented throughout the animal kingdom. The three known mammalian Hh proteins, Sonic (Shh), Desert (Dhh) and Indian (Ihh), are structurally related, and share a high degree of amino acid sequence identity (e.g. Shh and Ihh are 93% identical). The biologically active form of each Hh molecule is obtained by autocatalytic cleavage of their precursor proteins, and each corresponds to approximately one half of the N-terminal portion of the precursor molecule. Although Hh proteins have unique expression patterns and distinct biological roles within their respective regions of secretion, they use the same signaling pathway and can be substituted for one another in experimental systems. Recombinant Murine Sonic Hedgehog (Shh) is a 20.0 kDa protein consisting of 176 amino acid residues, including an N-terminal Ile-Val-Ile sequence substituted for the naturally occurring, chemically modified Cys residue.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.