Human Recombinant BAFF (from E. coli)
Supplier: Peprotech
310-13-5UG 310-13-20UG 310-13-100UG 310-13-250UG 310-13-500UG 310-13-1MG
10774-222EA 238.86 CAD
10774-222 10774-224 10774-226 10774-228 10774-230 10774-232
Human Recombinant BAFF (from E. coli)
Proteins and Peptides
BAFF, a member of the TNF superfamily of ligands, is expressed in T cells, macrophages, monocytes and dendritic cells. BAFF is involved in stimulation of B and T cell function, and is an important survival and maturation factor for peripheral B cells. BAFF signals through three different TNF receptors, TACI, BCMA and BAFF-R. The human BAFF gene codes for a 285 amino acid type II transmembrane protein containing a 46 amino acid cytoplasmic domain, a 21 amino acid transmembrane domain, and a 218 amino acid extracellular domain. Recombinant Human BAFF is a 152 amino acid polypeptide (17.0 kDa), which contains the TNF-like portion of the extracellular domain of BAFF.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.