Human Recombinant TFF-2 (from E. coli)
Supplier: Peprotech
The Trefoil Factor peptides (TFF1, TFF2 and TFF3) are expressed in the gastrointestinal tract, and appear to play an important role in intestinal mucosal defense and repair. TFF2 has been shown to inhibit gastrointestinal motility and gastric acid secretion. Recent data suggests a potential role for TFF2 in acute and chronic asthma (Nikolaidis, N.M. et al. Am. Journal Respir. Cell Mol. Biol. (2003) 4: 458-464). Recombinant Human TFF-2 is a 12.0 kDa polypeptide of 107 amino acid residues, which includes a 40-amino acid trefoil motif containing three conserved intramolecular disulfide bonds.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.