Human Recombinant LAG-1 (from E. coli)
Supplier: Peprotech
300-58-5UG 300-58-20UG 300-58-100UG 300-58-250UG 300-58-500UG 300-58-1MG
10774-026EA 221.22 CAD
10774-026 10774-028 10774-030 10774-032 10774-034 10774-036
Human Recombinant LAG-1 (from E. coli)
Proteins and Peptides
LAG-1 is CC chemokine that signals through the CCR5 receptor. LAG-1 is identical to MIP-1β (ACT II isotype) except for two amino acid substitutions; arginine for histidine at position 22 and serine for glycine at position 47 of the mature protein. LAG-1 chemoattracts monocytes, and exhibits activity as an HIV-suppressive factor. Recombinant Human LAG-1 (CCL4L1) is a 7.7 kDa protein containing 69 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.