Human Recombinant GCP-2 (from E. coli)
Supplier: Peprotech
300-41-5UG 300-41-20UG 300-41-100UG 300-41-250UG 300-41-500UG 300-41-1MG
10773-940EA 221.03 CAD
10773-940 10773-942 10773-944 10773-946 10773-948 10773-950
Human Recombinant GCP-2 (from E. coli)
Proteins and Peptides
CXCL6, also known as GCP-2 in humans, is a connective tissue-derived CXC chemokine that can signal through the CXCR1 and CXCR2 receptors. Human GCP-2, which is cross-reactive with murine cells, selectively attracts neutrophils, and has also been shown to exert anti-angiogenic activity. Recombinant Human GCP-2 (CXCL6) is a 7.9 kDa protein containing 72 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.