Human Recombinant HCC-1 (from E. coli)
Supplier: Peprotech
300-38-2UG 300-38-10UG 300-38-100UG 300-38-250UG 300-38-500UG 300-38-1MG
10773-908EA 209.84 CAD
10773-908 10773-910 10773-912 10773-914 10773-916 10773-918
Human Recombinant HCC-1 (from E. coli)
Proteins and Peptides
HCC-1 is a CC chemokine that signals through the CCR1 receptor and chemoattracts blood monocytes. It is secreted by various tissues including skeletal muscle, heart, spleen, liver and bone marrow. Mature HCC-1 is found in four different forms, which are distinguished by differential N-terminal truncation and contain 74, 72, 71, or 66 amino acid residues. Recombinant Human HCC-1 (CCL14) is an 8.4 kDa protein consisting of 72 amino acids, including the four highly conserved residues present in CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.