Human Recombinant TARC (from E. coli)
Supplier: PeproTech
300-30-100UG 300-30-1MG 300-30-500UG 300-30-20UG 300-30-5UG 300-30-250UG 300-30-50UG
10773-792EA 520.07 CAD
10773-792 10773-790 10773-802 10773-800 10773-798 10773-796 10773-794
Human Recombinant TARC (from E. coli)
Proteins and Peptides
TARC, a CC chemokine, is predominantly produced by dendritic cells in the thymus, and signals through the CCR4 receptor. TARC is chemotactic towards T cells. Recombinant Human TARC (CCL17) is an 8.0 kDa protein containing 71 amino acid residues, including the four conserved cysteine residues present in CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.