Human Recombinant SDF-1beta (from E. coli)
Supplier: Peprotech
300-28B-2UG 300-28B-10UG 300-28B-50UG 300-28B-100UG 300-28B-250UG 300-28B-500UG 300-28B-1MG
10773-752EA 209.84 CAD
10773-752 10773-754 10773-756 10773-758 10773-760 10773-762 10773-764
Human Recombinant SDF-1beta (from E. coli)
Proteins and Peptides
SDF-1α and β are stromal-derived, CXC chemokines that signal through the CXCR4 receptor. SDF-1α and β chemoattract B and T cells, and have been shown to induce migration of CD34+ stem cells. Additionally, the SDF-1 proteins exert HIV-suppressive activity in cells expressing the CXCR4 receptor. Human and murine SDF-1 proteins act across species. SDF-1α and β contain the four highly conserved cysteine residues present in CXC chemokines. The mature SDF-1β protein, produced by an N-terminal truncation of two additional amino acids, after removal of the signal sequence, contains 72 amino acid residues. Recombinant Human SDF-1β (CXCL12) is an 8.5 kDa protein containing 72 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.