Human Recombinant G-CSF (from E. coli)
Supplier: Peprotech
300-23-2UG 300-23-10UG 300-23-50UG 300-23-100UG 300-23-250UG 300-23-500UG 300-23-1MG
10773-678EA 223.76 CAD
10773-678 10773-680 10773-682 10773-684 10773-686 10773-688 10773-690
Human Recombinant G-CSF (from E. coli)
Proteins and Peptides
G-CSF is a hematopoietic growth factor that stimulates the development of committed progenitor cells to neutrophils and enhances the functional activities of the mature end-cell. It is produced in response to specific stimulation by a variety of cells, including macrophages, fibroblasts, endothelial cells and bone marrow stroma. G-CSF is being used clinically to facilitate hematopoietic recovery after bone marrow transplantation. Human and murine G-CSF are cross-species reactive. Recombinant Human G-CSF is an 18.7 kDa protein consisting of 174 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.