Human Recombinant Oncostatin (from E. coli)
Supplier: Peprotech
300-10T-2UG 300-10T-10UG 300-10T-100UG 300-10T-250UG 300-10T-500UG 300-10T-1MG
10773-530EA 223.76 CAD
10773-530 10773-532 10773-534 10773-536 10773-538 10773-540
Human Recombinant Oncostatin (from E. coli)
Proteins and Peptides
Oncostatin M (OSM) is a growth and differentiation factor that participates in the regulation of neurogenesis, osteogenesis and hematopoiesis. Produced by activated T cells, monocytes and Kaposi’s sarcoma cells, OSM can exert both stimulatory and inhibitory effects on cell proliferation. It stimulates the proliferation of fibroblasts, smooth muscle cells and Kaposi’s sarcoma cells, but inhibits the growth of some normal and tumor cell lines. It also promotes cytokine release (e.g. IL-6, GM-CSF and G-CSF) from endothelial cells, and enhances the expression of low-density lipoprotein receptors in hepatoma cells. OSM shares several structural and functional characteristics with LIF, IL-6, and CNTF. Human OSM is active on murine cells. The human OSM gene encodes for a 252 amino acid polypeptide, containing 25 amino acid signal sequence for secretion and a 227 precursor protein. Proteolytic processing of this precursor removes an 18 amino acid C-terminal peptide, and generates the mature OSM form. Recombinant Human Oncostatin M is a 23.6 kDa protein, containing 209 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.