Human Recombinant IFN-omega (from E. coli)
Supplier: Peprotech
300-02J-20UG 300-02J-100UG 300-02J-250UG
10773-500EA 226.59 CAD
10773-500 10773-502 10773-504
Human Recombinant IFN-omega (from E. coli)
Proteins and Peptides
IFN-ω is a type I interferon that can be induced by virus-infected leukocytes. Members of the type I interferon family, which includes IFN-α, IFN-β, and IFN-ω, signal through the IFNAR-1/IFNAR-2 receptor complex, and exert antiviral and antiproliferative activities. IFN-ω exhibits about 75% sequence homology with IFN-α, and contains two conserved disulfide bonds that are necessary for full biological activity. Recombinant Human IFN-ω is a 19.9 kDa protein consisting of 172 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.