Mouse Recombinant PF-4 (from E. coli)
Supplier: Peprotech
250-39-5UG 250-39-20UG 250-39-100UG 250-39-250UG 250-39-500UG 250-39-1MG
10773-438EA 209.84 CAD
10773-438 10773-440 10773-442 10773-444 10773-446 10773-448
Mouse Recombinant PF-4 (from E. coli)
Proteins and Peptides
PF-4 is a CXC chemokine that is expressed in megakaryocytes and stored in the α-granules of platelets. PF-4 is chemotactic towards neutrophils and monocytes, and has been shown to inhibit angiogenesis. Recombinant Murine PF-4 (CXCL4) is an 8.2 kDa protein containing 76 amino acid residues, including the four highly conserved residues present in CXC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.