Mouse Recombinant LIX (from E. coli)
Supplier: Peprotech
250-36-5UG 250-36-20UG 250-36-100UG 250-36-250UG 250-36-500UG 250-36-1MG
10773-402EA 209.84 CAD
10773-402 10773-404 10773-406 10773-408 10773-410 10773-412
Mouse Recombinant LIX (from E. coli)
Proteins and Peptides
CXCL6, also known as LIX in mice, is a CXC chemokine that signals through the CXCR2 receptor. It is expressed in monocytes, platelets, endothelial cells, and mast cells. LIX is a chemoattractant for neutrophils. The two naturally occurring variants of LIX, LIX 1-78 (GCP-2) and LIX 9-78 (GCP-2), contain 78 and 70 amino acid residues, respectively. LIX contains the four conserved cysteine residues present in CXC chemokines, and also contains the ‘ELR’ motif common to CXC chemokines that binds to the CXCR1 and CXCR2 receptors. Recombinant Murine LIX (CXCL6) is a 7.6 kDa protein consisting of amino acid residues 9-78 of the full length LIX protein (70 amino acids).
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.