Mouse Recombinant IFN-λ2 (from E. coli)
Supplier: Peprotech
250-33-100UG 250-33-250UG 250-33-500UG 250-33-1MG
10773-394EA 1566.84 CAD
10773-394 10773-396 10773-398 10773-400
Mouse Recombinant IFN-λ2 (from E. coli)
Proteins and Peptides
IFN λ1, 2, and 3 (also known as IL-29, IL-28A and IL-28B respectively) are distantly related to the IL-10 family and interferons. All three IFN-lambdas use a distinct receptor system composed of an IFN- λR1 subunit (also called CRF2-12) and IL-10R2 subunit (also called CRF2-14). Signaling through this receptor system induces antiviral defenses similar to, but distinct from, that of type I interferons. Recombinant Murine IFN- λ2 is a 19.8 kDa protein containing 175 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.