Mouse Recombinant CXCL16 (from E. coli)
Supplier: Peprotech
250-28-5UG 250-28-25UG 250-28-100UG 250-28-250UG 250-28-500UG 250-28-1MG
10773-382EA 233 CAD
10773-382 10773-384 10773-386 10773-388 10773-390 10773-392
Mouse Recombinant CXCL16 (from E. coli)
Proteins and Peptides
CXCL16 is a member of the CXC chemokine family and signals through the CXCR6 receptor. CXCL16 may play a role in attracting lymphocyte subsets during inflammation and may facilitate certain immune responses. The chemokine domain of CXCL16 contains six cysteine residues, including the four highly conserved cysteine residues characteristic of CXC chemokines. The CXCL16 gene codes for a 273 amino acid polypeptide, which includes a 29 amino acid cytoplasmic domain and transmembrane sequence containing approximately 20 amino acids. The extracellular portion of CXCL16 contains a chemokines domain and an extended C-terminal “mucin-like stalk” sequence. The extracellular domain contains 89 amino acid residues (86 a.a. residues for the murine homolog). Recombinant Murine CXCL16 is a 9.9 kDa protein containing 88 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.