Mouse Recombinant BCA-1/BLC (from E. coli)
Supplier: Peprotech
250-24-5UG 250-24-20UG 250-24-50UG 250-24-100UG 250-24-250UG 250-24-500UG 250-24-1MG
10773-316EA 220.96 CAD
10773-316 10773-318 10773-320 10773-322 10773-324 10773-326 10773-328
Mouse Recombinant BCA-1/BLC (from E. coli)
Proteins and Peptides
BCA-1/BLC, a CXC chemokine, is expressed in the liver, spleen, lymph nodes, appendix and stomach. It exerts its activities through its only receptor, CXCR5. BCA-1/BLC is a potent chemoattractant for B lymphocytes, and induces weak chemotactic response in T cells and macrophages. It manifests no activity on neutrophils and monocytes. Recombinant Murine BCA-1/BLC (CXCL13) is a 9.8 kDa protein containing 88 amino acid residues including the four highly conserved cysteine residues present in CXC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.