Mouse Recombinant MIG (from E. coli)
Supplier: Peprotech
250-18-5UG 250-18-20UG 250-18-100UG 250-18-250UG 250-18-500UG 250-18-1MG
10773-266EA 209.84 CAD
10773-266 10773-268 10773-270 10773-272 10780-280 10780-282
Mouse Recombinant MIG (from E. coli)
Proteins and Peptides
MIG, a CXC chemokine, is produced by IFN-γ stimulated monocytes, macrophages and endothelial cells. It signals through the CXCR3 receptor. MIG selectively chemoattracts Th1 lymphocytes, and also exerts other activities, including inhibition of tumor growth, angiogenesis, and inhibition of colony formation of hematopoietic progenitors. Human MIG is active on murine cells. Recombinant Murine MIG (CXCL9) is a 12.2 kDa protein containing 105 amino acid residues, including the four highly conserved cysteine residues present in CXC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.