Mouse Recombinant MIP-1gamma (from E. coli)
Supplier: Peprotech
MIP-1γ is a CC chemokine found in murine blood and a wide variety of murine tissues, with no known human homolog. MIP-1γ signals through the CCR1 receptor. MIP-1γ chemoattracts neutrophils, and also inhibits colony formation of bone marrow myeloid immature progenitors. MIP-1γ contains six cysteines, including the four highly conserved cysteine residues present in CC chemokines. Recombinant Murine MIP-1γ (CCL9/10) is an 11.6 kDa protein containing 101 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.