Mouse Recombinant KC (from E. coli)
Supplier: Peprotech
250-11-5UG 250-11-20UG 250-11-100UG 250-11-250UG 250-11-500UG 250-11-1MG
10773-180EA 223.76 CAD
10773-180 10773-182 10773-184 10773-186 10773-188 10773-190
Mouse Recombinant KC (from E. coli)
Proteins and Peptides
All three isoforms of GRO are CXC chemokines that can signal through the CXCR1 or CXCR2 receptors. The GRO proteins chemoattract and activate neutrophils and basophils. Recombinant Murine KC (CXCL1) is a 7.8 kDa protein consisting of 72 amino acids, including the 'ELR' motif common to the CXC chemokine family that binds to CXCR1 or CXCR2.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.