Human Recombinant IL-16 (from E. coli)
Supplier: Peprotech
200-16A-2UG 200-16A-10UG 200-16A-50UG 200-16A-100UG 200-16A-250UG 200-16A-500UG
10773-102EA 209.84 CAD
10773-102 10773-104 10773-106 10773-108 10773-110 10773-112
Human Recombinant IL-16 (from E. coli)
Proteins and Peptides
IL-16 is a CD8+ T cell-derived cytokine that induces chemotaxis of CD4+ T cells, CD4+ monocytes, and eosinophils. Analysis by gel filtration suggests that, under physiological conditions, hIL-16 exists predominantly as a noncovalently linked multimer, but that some IL-16 may exist as a monomer. However, only the multimeric form appears to possess chemotactic activity, suggesting that receptor cross-linking may be required for activity. IL-16 also induces expression of IL-2 receptor (IL-2R) and MHC class II molecules on CD4 + T cells. Human and murine IL-16 show significant cross-species reactivity. Recombinant Human IL-16 is a 12.4 kDa protein consisting of 121 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.