Human Recombinant IL-16 (from E. coli)
Supplier: Peprotech
200-16-2UG 200-16-10UG 200-16-50UG 200-16-100UG 200-16-250UG 200-16-500UG 200-16-1MG
10773-088EA 209.84 CAD
10773-088 10773-090 10773-092 10773-094 10773-096 10773-098 10773-100
Human Recombinant IL-16 (from E. coli)
Proteins and Peptides
IL-16 is a CD8+ T cell-derived cytokine that induces chemotaxis of CD4+ T cells, CD4+ monocytes, and eosinophils.
  • IL-16 also induces expression of IL-2 receptor (IL-2R) and MHC class II molecules on CD4 + T cells
  • Human and murine IL-16 show significant cross-species reactivity
  • Recombinant Human IL-16 is a 13.3 kDa protein consisting of 129 amino acid residues
Analysis by gel filtration suggests that, under physiological conditions, hIL-16 exists predominantly as a noncovalently linked multimer, but that some IL-16 may exist as a monomer. However, only the multimeric form appears to possess chemotactic activity, suggesting that receptor cross-linking may be required for activity.
Caution: For research use only. Not for use in diagnostic or therapeutic procedures.