Human Recombinant IL-15 (from E. coli)
Supplier: Peprotech
200-15-2UG 200-15-10UG 200-15-50UG 200-15-100UG 200-15-250UG 200-15-500UG 200-15-1MG
10773-074EA 241.19 CAD
10773-074 10773-076 10773-078 10773-080 10773-082 10773-084 10773-086
Human Recombinant IL-15 (from E. coli)
Proteins and Peptides
IL-15 is an immunomodulating cytokine that stimulates the proliferation of T lymphocytes and shares many biological properties with IL-2. IL-15 exerts its biological activities primarily on T cells. It is also essential in the development, survival and activation of NK cells. Increased expression of IL-15 has been linked to with rheumatoid arthritis, inflammatory bowel disease, and diseases affiliated with retroviruses HIV and HTLV-I. Human IL-15 is biologically active on mouse cells as measured by the dose-dependent stimulation of the proliferation of mouse CTLL cells. Recombinant Human IL-15 is a 12.9 kDa protein consisting of 115 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.