Human Recombinant IL-12 (from CHO cells)
Supplier: Peprotech
200-12-2UG 200-12-10UG 200-12-50UG 200-12-100UG 200-12-250UG 200-12-500UG 200-12-1MG
10773-010EA 233 CAD
10773-010 10773-012 10773-014 10773-016 10773-018 10773-020 10773-022
Human Recombinant IL-12 (from CHO cells)
Proteins and Peptides
IL-12 is a potent regulator of cell-mediated immune responses and it induces IFN-γ production by NK and T cells. It is produced by activated monocytes/macrophage cells, B lymphocytes and connective tissue-type mast cells. Among its biological activities, IL-12 promotes the growth and activity of activated NK, CD4+ and CD8+ cells, and induces the development of IFN-γ-producing Th1 cells. Recombinant Human IL-12 p70 is a 75.0 kDa heterodimeric glycoprotein consisting of disulfide-linked 35 kDa (p35) and 40 kDa (p40) subunits (503 total amino acid residues).
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.