Human Recombinant IL-10 (from E. coli)
Supplier: Peprotech
200-10-2UG 200-10-10UG 200-10-50UG 200-10-100UG 200-10-250UG 200-10-500UG 200-10-1MG
10772-984EA 228.75 CAD
10772-984 10772-986 10772-988 10772-990 10772-992 10772-994 10772-996
Human Recombinant IL-10 (from E. coli)
Proteins and Peptides
IL-10 is an immunosuppressive cytokine produced by a variety of mammalian cell types including macrophages, monocytes, T cells, B cells and keratinocytes. IL-10 inhibits the expression of proinflammatory cytokines such as IL-1 and TNF-α. Like IL-4, IL-10 enhances humoral immune responses and attenuates cell-mediated immune reactions. Human IL-10 is active on murine cells, but murine IL-10 is inactive on human cells. Recombinant Human IL-10 is an 18.6 kDa protein of 161 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.