Human Recombinant IL-9 (from E. coli)
Supplier: Peprotech
200-09-2UG 200-09-10UG 200-09-50UG 200-09-100UG 200-09-250UG 200-09-500UG 200-09-1MG
10772-970EA 239.65 CAD
10772-970 10772-972 10772-974 10772-976 10772-978 10772-980 10772-982
Human Recombinant IL-9 (from E. coli)
Proteins and Peptides
IL-9 is an immunoregulatory cytokine produced by IL-2 activated Th2 lymphocytes. IL-9 enhances the proliferation of T lymphocytes, mast cells, erythroid precursor cells and megakaryoblastic leukemia cell lines. Over-expression of IL-9 has been implicated in the pathogenesis of anaplastic lymphoma and Hodgkin’s disease. Whereas murine IL-9 can function on human cells, human IL-9 is inactive on mouse cells. Recombinant Human IL-9 is a 14.0 kDa protein of 127 amino acid residues, including 10 cysteine residues that are fully conserved between the human murine proteins.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.