Mouse Recombinant Granzyme (from Baculovirus (Insect cells))
Supplier: Peprotech
140-03-2UG 140-03-10UG
10772-830EA 209.84 CAD
10772-830 10772-832
Mouse Recombinant Granzyme (from Baculovirus (Insect cells))
Proteins and Peptides
Granzyme B is a cysteine protease found in the cytoplasmic granules of cytolytic T lymphocytes (CTL) and natural killer (NK) cells. Granzyme B is required for the induction of target cell lysis, which occurs as part of cell-mediated immune responses, and can activate apoptosis in target cells by both caspase-dependent and caspase-independent mechanisms. Proteolytic cleavage of substrates by Granzyme B takes place primarily after aspartic acid residues. Recombinant Murine Granzyme B is a glycosylated 227 amino acid protein, comprising the mature active portion of the murine Granzyme B precursor. The apparent molecular weight is 28.9 kDa by mass spectrometry.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.