Human Recombinant CTGFL/WISP-2 (from E. coli)
Supplier: Peprotech
120-16-5UG 120-16-20UG 120-16-100UG 120-16-250UG 120-16-500UG 120-16-1MG
10772-546EA 233 CAD
10772-546 10772-548 10772-550 10772-552 10772-554 10772-556
Human Recombinant CTGFL/WISP-2 (from E. coli)
Proteins and Peptides
CTGFL/WISP-2 is a 28.6 kDa protein that belongs to the CCN family of cysteine-rich regulatory proteins. Members of this family stimulate mitosis, adhesion, apoptosis, extracellular matrix production, growth arrest, and migration of multiple cell types. The protein is expressed in primary osteoblasts, fibroblasts, the ovaries, testes, and heart. In addition to promoting adhesion of osteoblasts, CTGFL/WISP-2 inhibits osteocalcin production, as well as binding of fibrinogen to integrin receptors. Mature human CTGFL/WISP-2 is a 24.8 kDa polypeptide protein containing 227 amino acids. It is composed of 3 distinct domains; the IGF-Binding Protein domain (IGF-BP), the Thrombospondin type I repeat (TSP type I), and von Willebrand Factor C motif (VWFC). Recombinant Human CTGFL/WISP-2 is a 24.3 kDa protein consisting of 228 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.