Human Recombinant TSG (from E. coli)

Supplier: Peprotech

120-09-10UG 120-09-50UG 120-09-100UG 120-09-250UG 120-09-500UG 120-09-1MG
10772-442EA 209.84 CAD
10772-442 10772-444 10772-446 10772-448 10772-450 10772-452
Human Recombinant TSG (from E. coli)
Proteins and Peptides

Twisted Gastrulation Protein (TSG) is a secreted BMP-binding protein that is structurally related to the BMP antagonists Chordin and Noggin. TSG can inhibit BMP activity by binding directly to BMP proteins, and can act either as a BMP-4 agonist or antagonist (depending on the specific biochemical environment) by binding to the BMP-4/Chordin complex. Recombinant Human TSG is a 198 amino acid, 22.2 kDa protein containing the BMP-4/TGF-β binding portion of the full length TSG protein.


Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.

Order Now

SPECIFICATIONS

Protein/peptide name TSG
Environmentally Preferable
Protein synonyms Twisted Gastrulation Protein
Protein/peptide type Recombinant
Protein Function Cytokines and Growth Factors
Species Human
Source E. coli
Conjugation Unconjugated
Gene ID 57045 Q9GZX9
Sequence CNKALCASDV SKCLIQELCQ CRPGEGNCSC CKECMLCLGA LWDECCDCVG MCNPRNYSDT PPTSKSTVEE LHEPIPSLFR ALTEGDTQLN WNIVSFPVAE ELSHHENLVS FLETVNQPHH QNVSVPSNNV HAPYSSDKEH MCTVVYFDDC MSIHQCKISC ESMGASKYRW FHNACCECIG PECIDYGSKT VKCMNCMF
Protease-free Y
Endotoxin level Low
Endotoxin content >0.1 ng/μg
Biological activity Biological Activity tests can vary. Please refer to the Product CoA for information.
Purity 0.95
Form Lyophilized
Formulation Formulation can vary. Please refer to product CoA for information.
Molecular weight 22.2 kDa
Reconstitution instructions Reconstitution instructions can vary. Please refer to the product CoA for info.
Storage conditions Store at –20 °C prior to reconstitution.
Shipping temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR