Human Recombinant FGF-16 (from E. coli)
Supplier: Peprotech
100-29-5UG 100-29-25UG 100-29-100UG 100-29-250UG 100-29-500UG 100-29-1MG
10772-140EA 227.33 CAD
10772-140 10772-142 10772-144 10772-146 10772-148 10772-150
Human Recombinant FGF-16 (from E. coli)
Proteins and Peptides
FGF-16 is a heparin-binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-16 signals through FGFR 2c and 3c. FGF-16 plays a role in the development of the central nervous system. Recombinant Human FGF-16 is a 23.6 kDa protein consisting of 206 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.