Human Recombinant FGF-10 (from E. coli)
Supplier: Peprotech
100-26-5UG 100-26-25UG 100-26-100UG 100-26-250UG 100-26-500UG 100-26-1MG
10772-104EA 234.5 CAD
10772-104 10772-106 10772-108 10772-110 10772-112 10772-114
Human Recombinant FGF-10 (from E. coli)
Proteins and Peptides
FGF-10 is a heparin-binding growth factor that belongs to the FGF family.
  • Recombinant Human FGF-10 is a 19.3 kDa protein consisting of 170 amino acid residues
Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-10 is most related to KGF/FGF-7, and is expressed during the development and, preferentially, in adult lungs. It signals through the FGFR 2b.
Caution: For research use only. Not for use in diagnostic or therapeutic procedures.