Human Recombinant VEGF-D (from HEK293 Cells)
Supplier: Peprotech
100-20D-2UG 100-20D-10UG 100-20D-100UG 100-20D-250UG 100-20D-500UG 100-20D-1MG
10772-022EA 226.59 CAD
10772-022 10772-024 10772-026 10772-028 10772-030 10772-032
Human Recombinant VEGF-D (from HEK293 Cells)
Proteins and Peptides
VEGF-D, a member of the VEGF/PDGF family of structurally related proteins, is a potent angiogenic cytokine. It promotes endothelial cell growth, promotes lymphangiogesis, and can also affect vascular permeability. VEGF-D is highly expressed in the lung, heart, small intestine and fetal lung, and at lower levels in the skeletal muscle, colon, and pancreas. It forms cell surface-associated, non-covalent, disulfide-linked homodimers, and can bind and activate both VEGFR-2 (flk1) and VEGFR-3 (flt4) receptors. During embryogenesis, VEGF-D may play a role in the formation of the venous and lymphatic vascular systems. It also participates in the growth and maintenance of differentiated lymphatic endothelium in adults. Both VEGF-C and VEGF-D are over-expressed in certain cancers, and the resulting elevated levels of VEGF-C or VEGF-D tend to correlate with increased lymphatic metastasis. Recombinant Human VEGF-D is a 26.2 kDa, non-disulfide linked, homodimeric protein consisting of two 117 amino acid polypeptide chains. Due to glycosylation, the protein migrates as a 20.0-22.0 kDa band by SDS-PAGE analysis under non-reducing conditions.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.