Human Recombinant KGF (from E. coli)
Supplier: Peprotech
100-19-2UG 100-19-10UG 100-19-50UG 100-19-100UG 100-19-250UG 100-19-500UG 100-19-1MG
10771-956EA 229.35 CAD
10771-956 10771-958 10771-960 10771-962 10771-964 10771-966 10771-968
Human Recombinant KGF (from E. coli)
Proteins and Peptides
KGF (FGF-7) is one of 23 known members of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth, and the regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. KGF (FGF-7) is a mitogen factor specific for epithelial cells and keratinocytes. KGF/FGF-7 signals through FGFR 2b. KGF (FGF-7) plays a role in kidney and lung development, as well as in angiogenesis and wound healing. Recombinant Human KGF (FGF-7) is an 18.9 kDa protein consisting of 163 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.