Rat Recombinant M-CSF (from E. coli)
Supplier: Peprotech
AF-400-28-2UG AF-400-28-10UG AF-400-28-50UG AF-400-28-100UG AF-400-28-250UG AF-400-28-500UG
10771-836EA 289.23 CAD
10771-836 10771-838 10771-840 10771-842 10771-844 10771-846
Rat Recombinant M-CSF (from E. coli)
Proteins and Peptides
M-CSF is a potent hematopoietic factor produced by a variety of cells, including lymphocytes, monocytes, fibroblasts, endothelial cells, myoblasts and osteoblasts. It is a key regulator of cellular proliferation, differentiation, and survival for blood monocytes, tissue macrophages, and their respective progenitor cells. M-CSF has been shown to play important roles in modulating dermal thickness and fertility. M-CSF is clinically used in the treatment of infection, malignancies and atherosclerosis. It facilitates hematopoietic recovery after bone marrow transplantation. Human M-CSF is reactive in murine systems, but the murine molecule exhibits no activity on human cells. Recombinant Rat M-CSF is a 36.2 kDa homodimeric protein consisting of two 155 amino acid polypeptide subunits.
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.