Rat Recombinant EGF (from E. coli)
Supplier: Peprotech
AF-400-25-20UG AF-400-25-100UG AF-400-25-250UG AF-400-25-500UG AF-400-25-1MG
10771-826EA 304.45 CAD
10771-826 10771-828 10771-830 10771-832 10771-834
Rat Recombinant EGF (from E. coli)
Proteins and Peptides
EGF is a potent growth factor that stimulates the proliferation of various epidermal and epithelial cells. Additionally, EGF has been shown to inhibit gastric secretion, and to be involved in wound healing. EGF signals through a receptor known as c-erbB, which is a class I tyrosine kinase receptor. This receptor also binds with TGF-α and VGF (vaccinia virus growth factor). Recombinant Rat EGF is a 6.2 kDa globular protein containing 54 amino acid residues, including 3 intramolecular disulfide bonds.
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.