Rat Recombinant SCF (from E. coli)
Supplier: Peprotech
AF-400-22-2UG AF-400-22-10UG AF-400-22-50UG AF-400-22-100UG AF-400-22-250UG AF-400-22-500UG AF-400-22-1MG
10771-798EA 289.23 CAD
10771-798 10771-800 10771-802 10771-804 10771-806 10771-808 10771-810
Rat Recombinant SCF (from E. coli)
Proteins and Peptides
SCF is a hematopoietic growth factor that exerts its activity by signaling through the c-Kit receptor. SCF and c-Kit are essential for the survival, proliferation and differentiation of hematopoietic cells committed to the melanocyte and germ cell lineages. Human SCF manifests low activity on murine cells, while murine and rat SCF are fully active on human cells. The human SCF gene encodes for a 273 amino acid transmembrane protein, which contains a 25 amino acid N-terminal signal sequence, a 189 amino acid extracellular domain, a 23 amino acid transmembrane domain, and a 36 amino acid cytoplasmic domain. The secreted soluble form of SCF is generated by proteolytic processing of the membrane anchored precursor. Recombinant Rat SCF is an 18.4 kDa polypeptide containing 165 amino acid residues, which corresponds to the sequence of the secreted soluble form of SCF.
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.