Mouse Recombinant Noggin (from E. coli)
Supplier: Peprotech
AF-250-38-5UG AF-250-38-20UG AF-250-38-50UG
10771-754EA 285.74 CAD
10771-754 10771-756 10771-758
Mouse Recombinant Noggin (from E. coli)
Proteins and Peptides
Noggin belongs to a group of diffusible proteins that bind to ligands of the TGF-β family, and regulate their activity by inhibiting their access to signaling receptors. Noggin was originally identified as a BMP-4 antagonist whose action was critical for proper formation of the head and other dorsal structures. Consequently, noggin has been shown to modulate the activities of other BMPs including BMP-2,-7,-13, and -14. Targeted deletion of noggin in mice results in prenatal death, and a recessive phenotype displaying a severely malformed skeletal system. Conversely, transgenic mice over-expressing noggin in mature osteoblasts display impaired osteoblastic differentiation, reduced bone formation, and severe osteoporosis. Recombinant Murine Noggin is a 46.4 kDa disulfide-linked homodimer consisting of two 206 amino acid polypeptide chains.
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.